Assay Details
Target Gene Details
Entrez Gene ID: | 51164 |
Gene Name: | dynactin subunit 4 |
Gene Aliases: |
DYN4, P62 |
Location: |
Chr.5:150708747-150759095 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCTN4 | NM_001135643.1 | 14 | 2091 | NP_001129115.1 |
| NM_001135644.1 | 13 | 2238 | NP_001129116.1 | |
| NM_016221.3 | 13 | 2070 | NP_057305.1 | |
| XM_005268457.1 | 12 | 1964 | XP_005268514.1 | |
| XM_011537644.1 | 14 | 1974 | XP_011535946.1 | |
| XM_011537645.1 | 14 | 1886 | XP_011535947.1 | |
| XM_011537646.1 | 14 | 2107 | XP_011535948.1 | |
| XM_011537647.1 | 14 | 1930 | XP_011535949.1 | |
| AF156165.2 | 1 | 462 | ||
| AF195120.1 | 13 | 1985 | AAF03896.1 | |
| AK000299.1 | 13 | 1989 | BAA91066.1 | |
| AK125973.1 | 13 | 2238 | ||
| AK296787.1 | ||||
| AL162085.1 | 3 | 1020 | CAB82417.1 | |
| BC026323.1 | 13 | 1995 | AAH26323.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759386 | Chr.5:150445030 - 150906180 on Build GRCh38 | Loss |
|
| nsv428129 | Chr.5:150641519 - 150906180 on Build GRCh38 | Loss |
|
| esv33898 | Chr.5:150657871 - 150766463 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map