Assay Details
Target Gene Details
Entrez Gene ID: | 9912 |
Gene Name: | Rho GTPase activating protein 44 |
Gene Aliases: |
NPC-A-10, RICH2 |
Location: |
Chr.17:12789512-12991644 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP44 | NM_001321164.1 | 1 | 315 | NP_001308093.1 |
| NM_001321166.1 | 1 | 315 | NP_001308095.1 | |
| NM_001321167.1 | 1 | 315 | NP_001308096.1 | |
| NM_001321168.1 | 1 | 315 | NP_001308097.1 | |
| NM_014859.5 | 1 | 315 | NP_055674.4 | |
| NR_135569.1 | 1 | 315 | ||
| XM_005256891.1 | 1 | 165 | XP_005256948.1 | |
| AB014572.1 | 1 | 288 | ||
| AK294538.1 | 1 | 60 | ||
| BC117412.1 | 1 | 67 | ||
| BC117416.1 | 1 | 67 | ||
| BC143853.1 | 1 | 67 | ||
| DA403021.1 | 1 | 287 | ||
| DB461035.1 | 1 | 318 | ||
| HY144351.1 | 1 | 309 |
Target Gene Details
Entrez Gene ID: | 100128006 |
Gene Name: | uncharacterized LOC100128006 |
Gene Aliases: |
- |
Location: |
Chr.17:12760134-12790284 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100128006 | NR_104607.1 | |||
| BX649083.1 | ||||
| DA231808.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34095 | Chr.17:12436261 - 12841981 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map