Assay Details
Target Gene Details
Entrez Gene ID: | 146050 |
Gene Name: | zinc finger and SCAN domain containing 29 |
Gene Aliases: |
ZNF690, Zfp690 |
Location: |
Chr.15:43358172-43371032 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZSCAN29 | NM_152455.3 | 3 | 980 | NP_689668.3 |
| XM_006720401.3 | 4 | 1740 | XP_006720464.1 | |
| XM_011521266.2 | 4 | 1420 | XP_011519568.1 | |
| XM_017021949.1 | 3 | 1952 | XP_016877438.1 | |
| AF525399.1 | 3 | 830 | AAO14995.1 | |
| AK093186.1 | 2 | 526 | BAC04088.1 | |
| AK093377.1 | BAC04147.1 | |||
| AK122788.1 | 3 | 968 | ||
| BC043612.1 | 3 | 948 | ||
| BC109270.1 | 3 | 830 | AAI09271.1 | |
| BC109271.1 | AAI09272.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1513 | Chr.15:43362170 - 43396516 on Build GRCh38 | Insertion |
|
| nsv1512 | Chr.15:43324636 - 43369448 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map