Assay Details
Target Gene Details
Entrez Gene ID: | 1840 |
Gene Name: | deltex E3 ubiquitin ligase 1 |
Gene Aliases: |
hDx-1 |
Location: |
Chr.12:113056711-113098028 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DTX1 | NM_004416.2 | 9 | 2165 | NP_004407.2 |
| XM_011538009.2 | 10 | 2807 | XP_011536311.1 | |
| AF053700.1 | AAC06246.1 | |||
| BC005816.2 | 5 | 644 | AAH05816.2 | |
| BC048216.1 | 9 | 2107 | AAH48216.1 |
Target Gene Details
Entrez Gene ID: | 8437 |
Gene Name: | RAS protein activator like 1 |
Gene Aliases: |
RASAL |
Location: |
Chr.12:113096515-113136239 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASAL1 | XM_005253950.4 | 22 | 5761 | XP_005254007.1 |
| XM_006719641.3 | 22 | 5600 | XP_006719704.1 | |
| XM_006719642.3 | 21 | 5730 | XP_006719705.1 | |
| XM_011538852.2 | 21 | 5733 | XP_011537154.1 | |
| XM_011538853.2 | 21 | 5727 | XP_011537155.1 | |
| XM_011538854.2 | 11 | 4473 | XP_011537156.1 | |
| XM_017020028.1 | 22 | 5758 | XP_016875517.1 | |
| XM_017020029.1 | 22 | 5843 | XP_016875518.1 | |
| XM_017020030.1 | 23 | 5871 | XP_016875519.1 | |
| XM_017020031.1 | 23 | 5868 | XP_016875520.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv560265 | Chr.12:113084969 - 113113365 on Build GRCh38 | Loss |
|
| nsv519540 | Chr.12:113042705 - 113459599 on Build GRCh38 | Gain+Loss |
|
| nsv560264 | Chr.12:113050137 - 113136469 on Build GRCh38 | Loss |
|
| esv3630792 | Chr.12:112988703 - 113838810 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map