Assay Details
Target Gene Details
Entrez Gene ID: | 1198 |
Gene Name: | CDC like kinase 3 |
Gene Aliases: |
PHCLK3, PHCLK3/152 |
Location: |
Chr.15:74598476-74645418 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLK3 | CR933693.1 | 4 | 2625 |
Target Gene Details
Entrez Gene ID: | 80153 |
Gene Name: | enhancer of mRNA decapping 3 |
Gene Aliases: |
LSM16, MRT50, YJDC, YJEFN2 |
Location: |
Chr.15:74630558-74696045 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EDC3 | NM_001142443.1 | NP_001135915.1 | ||
| NM_001142444.1 | NP_001135916.1 | |||
| NM_025083.3 | NP_079359.2 | |||
| XM_011522092.2 | XP_011520394.1 | |||
| AF193058.1 | AAG22486.1 | |||
| AK024781.1 | BAB15001.1 | |||
| AK056339.1 | ||||
| AK225099.1 | ||||
| BC011534.1 | AAH11534.1 | |||
| BC021271.2 | AAH21271.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv457204 | Chr.15:74592750 - 74655722 on Build GRCh38 | Loss |
|
| nsv817698 | Chr.15:74596822 - 74727108 on Build GRCh38 | Gain |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map