Assay Details
Target Gene Details
Entrez Gene ID: | 2935 |
Gene Name: | G1 to S phase transition 1 |
Gene Aliases: |
551G9.2, ETF3A, GST1, eRF3a |
Location: |
Chr.16:11868128-11916662 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GSPT1 | NM_001130006.1 | 15 | 2374 | NP_001123478.1 |
| NM_001130007.1 | 15 | 2398 | NP_001123479.1 | |
| NM_002094.3 | 15 | 2377 | NP_002085.2 | |
| AK095483.1 | 15 | 2305 | ||
| BC009503.2 | 15 | 2275 | AAH09503.2 | |
| X17644.1 | 15 | 2363 | CAA35635.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1043909 | Chr.16:11746054 - 11913578 on Build GRCh38 | Gain |
|
| nsv571458 | Chr.16:11831151 - 11993425 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map