Assay Details
Target Gene Details
Entrez Gene ID: | 57732 |
Gene Name: | zinc finger FYVE-type containing 28 |
Gene Aliases: |
LST2, LYST2 |
Location: |
Chr.4:2269585-2418698 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZFYVE28 | NM_001172656.1 | 12 | 3884 | NP_001166127.1 |
| NM_001172659.1 | 13 | 4007 | NP_001166130.1 | |
| NM_020972.2 | 13 | 3974 | NP_066023.2 | |
| XM_006713900.3 | 13 | 4338 | XP_006713963.1 | |
| XM_006713902.3 | 13 | 4704 | XP_006713965.1 | |
| XM_011513540.2 | 11 | 3752 | XP_011511842.1 | |
| XM_017008502.1 | 13 | 5777 | XP_016863991.1 | |
| XM_017008503.1 | 13 | 4707 | XP_016863992.1 | |
| XM_017008505.1 | 12 | 5572 | XP_016863994.1 | |
| AB046863.1 | 13 | 3973 | ||
| AK126692.1 | 12 | 4839 | ||
| BC041866.1 | 7 | 3675 | AAH41866.1 | |
| BM128958.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508990 | Chr.4:2246239 - 2324327 on Build GRCh38 | Insertion |
|
| nsv4200 | Chr.4:2252081 - 2286377 on Build GRCh38 | Insertion |
|
| nsv1009347 | Chr.4:2200935 - 2516063 on Build GRCh38 | Gain |
|
| esv2726835 | Chr.4:2019561 - 2959568 on Build GRCh38 | Deletion |
|
| nsv527189 | Chr.4:2243138 - 2276689 on Build GRCh38 | Loss |
|
| nsv829838 | Chr.4:2155167 - 2349950 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map