Assay Details
Target Gene Details
Entrez Gene ID: | 92747 |
Gene Name: | BPI fold containing family B member 1 |
Gene Aliases: |
C20orf114, LPLUNC1 |
Location: |
Chr.20:33283135-33309878 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BPIFB1 | NM_033197.2 | 1 | 49 | NP_149974.2 |
| AF364078.1 | AAM00283.1 | |||
| AK290243.1 | ||||
| AK310964.1 | ||||
| CD696225.1 | 1 | 71 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056411 | Chr.20:32469234 - 33355029 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map