Assay Details
Target Gene Details
Entrez Gene ID: | 3400 |
Gene Name: | inhibitor of DNA binding 4, HLH protein |
Gene Aliases: |
IDB4, bHLHb27 |
Location: |
Chr.6:19837370-19842200 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ID4 | NM_001546.3 | 3 | 2137 | NP_001537.1 |
| AJ420553.1 | 1 | 391 | ||
| AK130851.1 | 1 | 510 | ||
| BC014941.1 | 3 | 2121 | AAH14941.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv601090 | Chr.6:19840261 - 19859666 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs140771147] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map