Assay Details
Target Gene Details
Entrez Gene ID: | 6787 |
Gene Name: | NIMA related kinase 4 |
Gene Aliases: |
NRK2, STK2, pp12301 |
Location: |
Chr.3:52707936-52770949 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEK4 | NM_001193533.1 | 6 | 1220 | NP_001180462.1 |
| NM_003157.4 | 7 | 1487 | NP_003148.2 | |
| XM_006713310.2 | 7 | 1484 | XP_006713373.1 | |
| XM_011534039.2 | 7 | 1484 | XP_011532341.1 | |
| XM_011534040.2 | 7 | 1484 | XP_011532342.1 | |
| XM_017007085.1 | 7 | 1484 | XP_016862574.1 | |
| XM_017007086.1 | 7 | 1484 | XP_016862575.1 | |
| AK294165.1 | 6 | 1127 | ||
| AK312420.1 | 7 | 1475 | ||
| BC015515.1 | 7 | 1451 | ||
| BC063044.1 | 7 | 1477 | AAH63044.1 | |
| EF560744.1 | 7 | 1483 | ||
| KJ592714.1 | 7 | 1284 | ||
| L20321.1 | 7 | 1462 | AAA36658.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508219 | Chr.3:52707177 - 52769883 on Build GRCh38 | Deletion |
|
| esv2762328 | Chr.3:52740868 - 52968134 on Build GRCh38 | Loss |
|
| nsv834698 | Chr.3:52715967 - 52894203 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map