Assay Details
Target Gene Details
Entrez Gene ID: | 56985 |
Gene Name: | ADP-ribose/CDP-alcohol diphosphatase, manganese dependent |
Gene Aliases: |
C17orf48, MDS006, NBLA03831 |
Location: |
Chr.17:10697610-10711558 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADPRM | NM_020233.4 | 4 | 1400 | NP_064618.3 |
| DA280275.1 | 1 | 440 |
Target Gene Details
Entrez Gene ID: | 388335 |
Gene Name: | transmembrane protein 220 |
Gene Aliases: |
- |
Location: |
Chr.17:10711084-10730329 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM220 | XM_017024616.1 | 7 | 1462 | XP_016880105.1 |
| XM_017024617.1 | 6 | 1854 | XP_016880106.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2675560 | Chr.17:10710661 - 10748164 on Build GRCh38 | Deletion |
|
| nsv1065819 | Chr.17:10622619 - 10784575 on Build GRCh38 | Gain |
|
| esv3639941 | Chr.17:10710674 - 10748133 on Build GRCh38 | Loss |
|
| esv3639942 | Chr.17:10710767 - 10713237 on Build GRCh38 | Loss |
|
| nsv1063900 | Chr.17:10662646 - 10729182 on Build GRCh38 | Gain |
|
| nsv833367 | Chr.17:10683841 - 10826633 on Build GRCh38 | Loss |
|
| nsv1058835 | Chr.17:10322963 - 10939602 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map