Assay Details
Target Gene Details
Entrez Gene ID: | 7014 |
Gene Name: | telomeric repeat binding factor 2 |
Gene Aliases: |
TRBF2, TRF2 |
Location: |
Chr.16:69355561-69386004 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TERF2 | NM_005652.4 | NP_005643.2 | ||
| XM_005256121.3 | XP_005256178.1 | |||
| XM_005256122.3 | XP_005256179.1 | |||
| XM_005256123.3 | XP_005256180.1 | |||
| XM_005256124.3 | XP_005256181.1 | |||
| AF002999.1 | AAB81135.1 | |||
| U95970.1 | AAD00821.1 | |||
| X93512.1 | CAA63769.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv974826 | Chr.16:69357471 - 69358853 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map