Assay Details
Target Gene Details
Entrez Gene ID: | 29925 |
Gene Name: | GDP-mannose pyrophosphorylase B |
Gene Aliases: |
MDDGA14, MDDGB14, MDDGC14 |
Location: |
Chr.3:49721476-49723974 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GMPPB | NM_013334.3 | 5 | 768 | NP_037466.2 |
| NM_021971.2 | 5 | 768 | NP_068806.1 | |
| AF135421.1 | 5 | 660 | AAD38516.1 | |
| AK024319.1 | 5 | 745 | BAB14882.1 | |
| AK291700.1 | 5 | 743 | ||
| BC001141.1 | 5 | 707 | AAH01141.1 | |
| BC008033.1 | 5 | 707 | AAH08033.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508217 | Chr.3:49578013 - 49732463 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map