Assay Details
Target Gene Details
Entrez Gene ID: | 6102 |
Gene Name: | retinitis pigmentosa 2 (X-linked recessive) |
Gene Aliases: |
DELXp11.3, NM23-H10, NME10, TBCCD2, XRP2 |
Location: |
Chr.X:46836912-46882358 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RP2 | NM_006915.2 | 5 | 2413 | NP_008846.2 |
| AJ007590.1 | 5 | 2396 | CAA07577.1 | |
| AL049366.1 | ||||
| BC043348.2 | 5 | 2335 | AAH43348.1 | |
| BC053530.1 | 5 | 2333 | AAH53530.1 |
More Information
Set Membership: |
|

Genomic Map