Assay Details
Target Gene Details
Entrez Gene ID: | 5993 |
Gene Name: | regulatory factor X5 |
Gene Aliases: |
- |
Location: |
Chr.1:151340640-151347319 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RFX5 | NM_000449.3 | 3 | 252 | NP_000440.1 |
| NM_001025603.1 | 3 | 245 | NP_001020774.1 | |
| XM_005245405.1 | 3 | 307 | XP_005245462.1 | |
| XM_005245406.3 | 3 | 264 | XP_005245463.1 | |
| XM_011509847.1 | 3 | 311 | XP_011508149.1 | |
| XM_011509848.1 | 3 | 314 | XP_011508150.1 | |
| XM_011509849.1 | 3 | 247 | XP_011508151.1 | |
| XM_011509850.1 | 3 | 374 | XP_011508152.1 | |
| XM_017001999.1 | 3 | 307 | XP_016857488.1 | |
| XM_017002000.1 | 3 | 338 | XP_016857489.1 | |
| AK223526.1 | 1 | 44 | BAD97246.1 | |
| AK302891.1 | 3 | 245 | ||
| AK309072.1 | 3 | 264 | ||
| AK311525.1 | 3 | 249 | ||
| AK312289.1 | 3 | 225 | ||
| BC017471.1 | 3 | 179 | AAH17471.1 | |
| BX423384.2 | 3 | 278 | ||
| X85786.1 | 2 | 192 | CAA59771.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522840 | Chr.1:150961247 - 151375270 on Build GRCh38 | Gain |
|
| esv34461 | Chr.1:151268066 - 151439159 on Build GRCh38 | Gain |
|
| nsv438026 | Chr.1:151346257 - 151372227 on Build GRCh38 | Loss |
|
| nsv831581 | Chr.1:151316675 - 151506129 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map