Assay Details
Target Gene Details
Entrez Gene ID: | 2729 |
Gene Name: | glutamate-cysteine ligase catalytic subunit |
Gene Aliases: |
GCL, GCS, GLCL, GLCLC |
Location: |
Chr.6:53497341-53545129 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCLC | NM_001197115.1 | 15 | 2282 | NP_001184044.1 |
| NM_001498.3 | 16 | 2396 | NP_001489.1 | |
| XM_017010749.1 | 16 | 2021 | XP_016866238.1 | |
| AB262176.1 | 16 | 2299 | ||
| AB516284.1 | ||||
| AK094940.1 | 1 | 1433 | ||
| AK304288.1 | 16 | 1924 | ||
| AK313480.1 | ||||
| BC039894.1 | 16 | 1994 | AAH39894.1 | |
| M90656.1 | 16 | 2004 | AAA58499.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763940 | Chr.6:53228517 - 53622284 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map