Assay Details
Target Gene Details
Entrez Gene ID: | 79632 |
Gene Name: | family with sequence similarity 184 member A |
Gene Aliases: |
C6orf60 |
Location: |
Chr.6:118959829-119149193 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM184A | NM_001100411.2 | NP_001093881.1 | ||
| NM_001288576.1 | NP_001275505.1 | |||
| NM_024581.5 | NP_078857.5 | |||
| AK024004.1 | BAB14769.1 | |||
| AK091682.1 | BAC03719.1 | |||
| AK127744.1 | BAC87110.1 | |||
| AL136767.1 | CAB66701.1 | |||
| AY131226.1 | AAN07912.1 | |||
| BC009055.1 | AAH09055.2 | |||
| BC060769.1 | AAH60769.2 | |||
| BX538186.1 | CAD98058.1 |
More Information
Set Membership: |
|

Genomic Map