Assay Details
Target Gene Details
Entrez Gene ID: | 57175 |
Gene Name: | coronin 1B |
Gene Aliases: |
CORONIN-2 |
Location: |
Chr.11:67438047-67443821 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CORO1B | NM_001018070.2 | 11 | 1274 | NP_001018080.1 |
| NM_020441.2 | 10 | 1252 | NP_065174.1 | |
| AK000860.1 | 10 | 1235 | ||
| AK315399.1 | 10 | 1207 | ||
| AL162072.1 | 5 | 525 | CAB82408.1 | |
| BC006449.1 | 10 | 1202 | AAH06449.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv468604 | Chr.11:67418529 - 67516420 on Build GRCh38 | Loss |
|
| dgv1977n54 | Chr.11:67397024 - 67521123 on Build GRCh38 | Loss |
|
| dgv52n111 | Chr.11:67362651 - 67521123 on Build GRCh38 | Deletion |
|
| nsv521723 | Chr.11:67408221 - 67543787 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map