Assay Details
Target Gene Details
Entrez Gene ID: | 79837 |
Gene Name: | phosphatidylinositol-5-phosphate 4-kinase type 2 gamma |
Gene Aliases: |
PIP5K2C |
Location: |
Chr.12:57591159-57603428 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIP4K2C | NM_001146258.1 | NP_001139730.1 | ||
| NM_001146259.1 | 10 | 1389 | NP_001139731.1 | |
| NM_001146260.1 | 9 | 1299 | NP_001139732.1 | |
| NM_024779.4 | 10 | 1443 | NP_079055.3 | |
| XM_005269152.2 | 9 | 1397 | XP_005269209.1 | |
| XM_017019973.1 | 11 | 1505 | XP_016875462.1 | |
| AK025708.1 | 6 | 722 | BAB15223.1 | |
| AK075553.1 | ||||
| AK125526.1 | 10 | 1327 | ||
| AK225343.1 | 6 | 722 | ||
| AK225685.1 | 6 | 730 | ||
| AK297243.1 | 9 | 1284 | ||
| AK302254.1 | 10 | 1305 | ||
| AX754970.1 | 10 | 1384 | CAD99241.1 | |
| BC028596.2 | 10 | 1328 | AAH28596.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428283 | Chr.12:57435950 - 57608085 on Build GRCh38 | Gain |
|
| nsv559034 | Chr.12:57466149 - 57618450 on Build GRCh38 | Loss |
|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map