Assay Details
Target Gene Details
Entrez Gene ID: | 80122 |
Gene Name: | mitogen-activated protein kinase kinase kinase 19 |
Gene Aliases: |
RCK, YSK4 |
Location: |
Chr.2:134963875-135047468 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
More Information
Set Membership: |
|

Genomic Map