Assay Details
Target Gene Details
Entrez Gene ID: | 51438 |
Gene Name: | MAGE family member C2 |
Gene Aliases: |
CT10, HCA587, MAGEE1 |
Location: |
Chr.X:142202342-142205290 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEC2 | NM_016249.3 | NP_057333.1 | ||
| AF116194.1 | AAF34816.1 | |||
| AF151378.1 | AAF36533.1 | |||
| AF196482.1 | AAF07210.1 | |||
| AK313591.1 | ||||
| BC013318.1 | AAH13318.1 | |||
| BP370466.1 |
More Information
Set Membership: |
|

Genomic Map