Assay Details
Target Gene Details
Entrez Gene ID: | 84896 |
Gene Name: | ATPase family, AAA domain containing 1 |
Gene Aliases: |
AFDC1, FNP001, THORASE |
Location: |
Chr.10:87751512-87841345 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATAD1 | NM_001321967.1 | NP_001308896.1 | ||
| NM_001321968.1 | NP_001308897.1 | |||
| NM_001321969.1 | NP_001308898.1 | |||
| NM_032810.3 | NP_116199.2 | |||
| NR_135914.1 | ||||
| XM_005270252.4 | XP_005270309.1 | |||
| XM_011540302.1 | XP_011538604.1 | |||
| XM_017016847.1 | XP_016872336.1 | |||
| XM_017016848.1 | XP_016872337.1 | |||
| XM_017016849.1 | XP_016872338.1 | |||
| XM_017016850.1 | XP_016872339.1 | |||
| AF361493.1 | AAL57218.1 | |||
| AK027506.1 | BAB55161.1 | |||
| AK075223.1 | BAC11482.1 | |||
| AK304297.1 | ||||
| AL834370.1 | CAD39033.1 | |||
| BC010868.1 | AAH10868.1 | |||
| BC063530.1 | AAH63530.1 | |||
| BC073998.1 | AAH73998.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518233 | Chr.10:87753082 - 87824245 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map