Assay Details
Target Gene Details
Entrez Gene ID: | 119687 |
Gene Name: | olfactory receptor family 51 subfamily A member 7 |
Gene Aliases: |
OR11-27 |
Location: |
Chr.11:4907370-4908308 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR51A7 | NM_001004749.1 | 1 | 882 | NP_001004749.1 |
| BC137062.1 | 1 | 919 | ||
| BC137063.1 | 1 | 919 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891914 | Chr.11:4642313 - 4974456 on Build GRCh38 | Gain |
|
| nsv1054847 | Chr.11:4621645 - 5179426 on Build GRCh38 | Gain |
|
| esv2759799 | Chr.11:4886663 - 5114101 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map