Assay Details
Target Gene Details
Entrez Gene ID: | 9947 |
Gene Name: | MAGE family member C1 |
Gene Aliases: |
CT7, CT7.1 |
Location: |
Chr.X:141903856-141909401 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEC1 | NM_005462.4 | NP_005453.2 | ||
| XM_011531418.2 | XP_011529720.1 | |||
| DB065460.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs146483199] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map