Assay Details
Target Gene Details
Entrez Gene ID: | 552 |
Gene Name: | arginine vasopressin receptor 1A |
Gene Aliases: |
AVPR V1a, AVPR1, V1aR |
Location: |
Chr.12:63142759-63152810 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AVPR1A | NM_000706.4 | 2 | 3429 | NP_000697.1 |
| L25615.1 | AAA62271.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469592 | Chr.12:63092730 - 63285359 on Build GRCh38 | Loss |
|
| esv2761742 | Chr.12:63143632 - 63188553 on Build GRCh38 | Loss |
|
| nsv832436 | Chr.12:63110213 - 63264332 on Build GRCh38 | Gain |
|
| nsv1046836 | Chr.12:62998611 - 63983389 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map