Assay Details
Target Gene Details
Entrez Gene ID: | 9703 |
Gene Name: | KIAA0100 |
Gene Aliases: |
BCOX, BCOX1, CT101, FMP27 |
Location: |
Chr.17:28614440-28645159 on Build GRCh38 |
Assay Gene Location: | Within Exon 39 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIAA0100 | NM_014680.3 | 39 | 7035 | NP_055495.2 |
| XM_005258073.2 | 39 | 7027 | XP_005258130.1 | |
| XM_017025423.1 | 38 | 6884 | XP_016880912.1 | |
| BC008591.2 | 9 | 1339 | AAH08591.2 | |
| BC103726.1 | 9 | 1461 | AAI03727.1 | |
| BC120873.1 | ||||
| BM999622.1 | 1 | 418 | ||
| CN482281.1 | 1 | 194 | ||
| CR933634.1 | 40 | 6894 | CAI45940.1 | |
| D43947.3 | 39 | 7007 | BAA07891.3 |
Target Gene Details
Entrez Gene ID: | 100506436 |
Gene Name: | SPAG5 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:28599133-28617377 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPAG5-AS1 | NR_040012.1 | |||
| AK098638.1 | ||||
| DB457167.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064087 | Chr.17:28462253 - 28755692 on Build GRCh38 | Gain |
|

Genomic Map