Assay Details
Target Gene Details
Entrez Gene ID: | 6016 |
Gene Name: | Ras like without CAAX 1 |
Gene Aliases: |
NS8, RIBB, RIT, ROC1 |
Location: |
Chr.1:155897808-155911402 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RIT1 | NM_001256820.1 | 5 | 792 | NP_001243749.1 |
| NM_001256821.1 | 6 | 819 | NP_001243750.1 | |
| NM_006912.5 | 6 | 941 | NP_008843.1 | |
| AF084462.1 | AAD13021.1 | |||
| AK298768.1 | 5 | 792 | ||
| BC104186.2 | 6 | 812 | AAI04187.1 | |
| BC104187.2 | 6 | 812 | AAI04188.1 | |
| BM546985.1 | 6 | 935 | ||
| U71203.1 | 6 | 873 | AAB42213.1 | |
| U78165.1 | 6 | 873 | AAB64246.1 | |
| Y07566.1 | 6 | 883 | CAA68851.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33869 | Chr.1:155253492 - 155948170 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map