Assay Details
Target Gene Details
Entrez Gene ID: | 163782 |
Gene Name: | KN motif and ankyrin repeat domains 4 |
Gene Aliases: |
ANKRD38, dJ1078M7.1 |
Location: |
Chr.1:62236165-62319414 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KANK4 | NM_001320269.1 | 9 | 2044 | NP_001307198.1 |
| NM_181712.4 | 10 | 3926 | NP_859063.3 | |
| XM_011540843.2 | 10 | 3811 | XP_011539145.1 | |
| XM_011540845.2 | 10 | 3805 | XP_011539147.1 | |
| XM_017000483.1 | 9 | 3675 | XP_016855972.1 | |
| XM_017000484.1 | 10 | 2180 | XP_016855973.1 | |
| XM_017000485.1 | 8 | 2112 | XP_016855974.1 | |
| AB284127.1 | 10 | 3926 | ||
| AK091941.1 | 9 | 1931 | BAC03774.1 | |
| AK096259.1 | 4 | 1310 | BAC04741.1 | |
| AK127461.1 | 7 | 2852 | ||
| AL832644.2 | 8 | 2678 | CAD89966.1 | |
| BC041418.1 | 8 | 2146 | AAH41418.1 | |
| BC060866.1 | 10 | 3926 | AAH60866.1 | |
| BU738703.1 | 1 | 586 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893601 | Chr.1:62162003 - 62343094 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map