Assay Details
Target Gene Details
Entrez Gene ID: | 246243 |
Gene Name: | ribonuclease H1 |
Gene Aliases: |
H1RNA, PEOB2, RNH1 |
Location: |
Chr.2:3531813-3558371 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNASEH1 | NM_001286834.1 | NP_001273763.1 | ||
| NM_001286837.1 | NP_001273766.1 | |||
| NM_002936.4 | NP_002927.2 | |||
| AF039652.1 | AAC09261.1 | |||
| AF048994.1 | AAC78563.1 | |||
| AF048995.1 | AAC78564.1 | |||
| AJ224117.1 | CAA11835.1 | |||
| AK075490.1 | ||||
| AK096913.1 | ||||
| BC002973.1 | AAH02973.1 | |||
| BT019670.1 | AAV38476.1 | |||
| CR541820.1 | CAG46619.1 | |||
| DA532761.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819972 | Chr.2:3551407 - 3560402 on Build GRCh38 | Gain |
|
| nsv1011656 | Chr.2:2931099 - 3576389 on Build GRCh38 | Gain |
|
| dgv3702n100 | Chr.2:3361936 - 3733306 on Build GRCh38 | Gain |
|
| nsv580822 | Chr.2:3552110 - 4305551 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map