Assay Details
Target Gene Details
Entrez Gene ID: | 7812 |
Gene Name: | cold shock domain containing E1 |
Gene Aliases: |
D1S155E, UNR |
Location: |
Chr.1:114716913-114758050 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSDE1 | NM_001007553.2 | 20 | 3850 | NP_001007554.1 |
| NM_001130523.2 | 20 | 3904 | NP_001123995.1 | |
| NM_001242891.1 | 21 | 3997 | NP_001229820.1 | |
| NM_001242892.1 | 19 | 3463 | NP_001229821.1 | |
| NM_001242893.1 | 18 | 3370 | NP_001229822.1 | |
| NM_007158.5 | 19 | 3757 | NP_009089.4 | |
| AB020692.1 | 20 | 3764 | ||
| BU681634.1 | 1 | 334 | ||
| BX648780.1 | 19 | 3691 | ||
| CR749378.1 | 20 | 3854 | CAH18231.1 | |
| CR749431.1 | 20 | 3836 | CAH18269.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv28734 | Chr.1:114376708 - 115005298 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map