Assay Details
Target Gene Details
Entrez Gene ID: | 3845 |
Gene Name: | KRAS proto-oncogene, GTPase |
Gene Aliases: |
C-K-RAS, CFC2, K-RAS2A, K-RAS2B, K-RAS4A, K-RAS4B, KI-RAS, KRAS1, KRAS2, NS, NS3, RALD, RASK2 |
Location: |
Chr.12:25204789-25252093 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRAS | NM_004985.4 | 5 | 3513 | NP_004976.2 |
| NM_033360.3 | 6 | 3637 | NP_203524.1 | |
| XM_011520653.2 | 5 | 3505 | XP_011518955.1 | |
| XM_017019293.1 | 6 | 4125 | XP_016874782.1 | |
| CD245555.1 | 1 | 309 | ||
| M54968.1 | 5 | 3513 | AAB41942.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv557798 | Chr.12:25206907 - 25244348 on Build GRCh38 | Loss |
|
| nsv1048631 | Chr.12:25200583 - 25225727 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map