Assay Details
Target Gene Details
Entrez Gene ID: | 158809 |
Gene Name: | MAGE family member B6 |
Gene Aliases: |
CT3.4, MAGE-B6, MAGEB6A |
Location: |
Chr.X:26192440-26195646 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEB6 | NM_173523.2 | 2 | 699 | NP_775794.2 |
| AK097561.1 | 2 | 699 | BAC05102.1 | |
| BC067286.1 | 2 | 699 | AAH67286.1 | |
| BC074920.2 | 1 | 587 | AAH74920.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2740023 | Chr.X:26193893 - 26194423 on Build GRCh38 | Deletion |
|
| esv2740021 | Chr.X:26165089 - 26198461 on Build GRCh38 | Deletion |
|
| esv2740022 | Chr.X:26193603 - 26194624 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs145782491] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map