Assay Details
Target Gene Details
Entrez Gene ID: | 6780 |
Gene Name: | staufen double-stranded RNA binding protein 1 |
Gene Aliases: |
PPP1R150, STAU |
Location: |
Chr.20:49113339-49219295 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STAU1 | NM_001037328.2 | 14 | 3411 | NP_001032405.1 |
| NM_001319134.1 | 13 | 3288 | NP_001306063.1 | |
| NM_001319135.1 | 15 | 3470 | NP_001306064.1 | |
| NM_001322927.1 | 12 | 3213 | NP_001309856.1 | |
| NM_001322928.1 | 15 | 3557 | NP_001309857.1 | |
| NM_001322929.1 | 15 | 3682 | NP_001309858.1 | |
| NM_001322930.1 | 13 | 3484 | NP_001309859.1 | |
| NM_001322931.1 | 16 | 3662 | NP_001309860.1 | |
| NM_001322932.1 | 13 | 3502 | NP_001309861.1 | |
| NM_001322933.1 | 14 | 3577 | NP_001309862.1 | |
| NM_004602.3 | 14 | 3393 | NP_004593.2 | |
| NM_017452.3 | 13 | 3270 | NP_059346.2 | |
| NM_017453.3 | 14 | 3559 | NP_059347.2 | |
| NM_017454.3 | 12 | 3195 | NP_059348.2 | |
| XM_005260527.1 | 14 | 3460 | XP_005260584.1 | |
| XM_006723865.1 | 15 | 3697 | XP_006723928.1 | |
| XM_011529015.1 | 16 | 3810 | XP_011527317.1 | |
| XM_017028028.1 | 15 | 3536 | XP_016883517.1 | |
| AJ132258.1 | 11 | 2937 | CAB40082.1 | |
| AK091405.1 | 10 | 2749 | ||
| AY529074.1 | AAS76636.1 | |||
| AY546099.1 | AAS76637.1 | |||
| BE439382.1 | 1 | 149 | ||
| BQ649508.1 | 1 | 796 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3646010 | Chr.20:49101246 - 49121848 on Build GRCh38 | Gain |
|
| nsv1067146 | Chr.20:48878894 - 49475527 on Build GRCh38 | Loss |
|
| esv2762079 | Chr.20:49095851 - 49155612 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map