Assay Details
Target Gene Details
Entrez Gene ID: | 283159 |
Gene Name: | olfactory receptor family 8 subfamily D member 1 |
Gene Aliases: |
JCG9, OR8D3, OST004, PDJ9J14 |
Location: |
Chr.11:124309840-124310766 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR8D1 | NM_001002917.1 | 1 | 440 | NP_001002917.1 |
| AF238489.1 | 1 | 440 | AAL32998.1 | |
| BC136950.1 | 1 | 511 | ||
| BC136954.1 | 1 | 511 |
Target Gene Details
Entrez Gene ID: | 219865 |
Gene Name: | olfactory receptor family 8 subfamily G member 5 |
Gene Aliases: |
OR11-298, OR8G5P, OR8G6 |
Location: |
Chr.11:124241158-124323283 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR8G5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv826113 | Chr.11:124267376 - 124372263 on Build GRCh38 | Loss |
|
| esv3892105 | Chr.11:124309052 - 124530715 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map