Assay Details
Target Gene Details
Entrez Gene ID: | 54930 |
Gene Name: | HAUS augmin like complex subunit 4 |
Gene Aliases: |
C14orf94 |
Location: |
Chr.14:22946228-22957142 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HAUS4 | NM_001166269.1 | NP_001159741.1 | ||
| NM_001166270.1 | NP_001159742.1 | |||
| NM_017815.2 | NP_060285.2 | |||
| AK000431.1 | BAA91161.1 | |||
| AK026016.1 | BAB15322.1 | |||
| AK057779.1 | ||||
| AY927599.1 | ||||
| BC001916.1 | AAH01916.1 | |||
| BC002554.2 | AAH02554.1 | |||
| BX247981.1 | CAD62315.1 | |||
| BX247995.1 | CAD62328.1 | |||
| BX248256.1 | CAD62584.1 |
Target Gene Details
Entrez Gene ID: | 101926933 |
Gene Name: | uncharacterized LOC101926933 |
Gene Aliases: |
- |
Location: |
Chr.14:22929609-22954692 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101926933 | NR_110002.1 | |||
| DA905235.1 |
More Information
Set Membership: |
|

Genomic Map