Assay Details
Target Gene Details
Entrez Gene ID: | 784 |
Gene Name: | calcium voltage-gated channel auxiliary subunit beta 3 |
Gene Aliases: |
CAB3, CACNLB3 |
Location: |
Chr.12:48814432-48828943 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNB3 | NM_000725.3 | NP_000716.2 | ||
| NM_001206915.1 | NP_001193844.1 | |||
| NM_001206916.1 | NP_001193845.1 | |||
| NM_001206917.1 | NP_001193846.1 | |||
| XM_011538727.2 | XP_011537029.1 | |||
| XM_011538728.2 | XP_011537030.1 | |||
| XM_017019957.1 | XP_016875446.1 | |||
| XM_017019958.1 | XP_016875447.1 | |||
| XM_017019959.1 | XP_016875448.1 | |||
| AK122911.1 | ||||
| AK289709.1 | ||||
| AK297639.1 | ||||
| AK300901.1 | ||||
| AK304537.1 | ||||
| AK316323.1 | ||||
| AL534844.3 | ||||
| AL537072.3 | ||||
| AL702978.1 | ||||
| BC041811.1 | AAH41811.1 | |||
| BI667991.1 | ||||
| CD013906.1 | ||||
| L27584.1 | AAA19799.1 | |||
| U07139.1 | AAA95958.1 | |||
| X76555.1 | CAA54055.1 | |||
| X76556.1 | CAA54056.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv698 | Chr.12:48795148 - 48828796 on Build GRCh38 | Insertion |
|
| nsv826363 | Chr.12:48783444 - 48857206 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map