Assay Details
Target Gene Details
Entrez Gene ID: | 285381 |
Gene Name: | diphthamide biosynthesis 3 |
Gene Aliases: |
DELGIP, DELGIP1, DESR1, DPH3A, KTI11, ZCSL2 |
Location: |
Chr.3:16257061-16264989 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPH3 | NM_001047434.2 | NP_001040899.1 | ||
| NM_206831.2 | NP_996662.1 | |||
| AK022970.1 | ||||
| AK092040.1 | ||||
| AK311618.1 | ||||
| BC010181.2 | AAH10181.1 | |||
| DA447430.1 | ||||
| DA875039.1 |
More Information
Set Membership: |
|

Genomic Map