Assay Details
Target Gene Details
Entrez Gene ID: | 54443 |
Gene Name: | anillin actin binding protein |
Gene Aliases: |
FSGS8, Scraps, scra |
Location: |
Chr.7:36389803-36453791 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANLN | NM_001284301.2 | NP_001271230.1 | ||
| NM_001284302.2 | NP_001271231.1 | |||
| NM_018685.4 | NP_061155.2 | |||
| XM_006715746.1 | XP_006715809.1 | |||
| XM_006715747.3 | XP_006715810.1 | |||
| XM_017012354.1 | XP_016867843.1 | |||
| XM_017012355.1 | XP_016867844.1 | |||
| XM_017012356.1 | XP_016867845.1 | |||
| AF273437.1 | AAF75796.1 | |||
| AK001468.1 | BAA91710.1 | |||
| AK001472.1 | BAA91711.1 | |||
| AK023208.1 | BAB14463.1 | |||
| AK291254.1 | ||||
| AK299803.1 | ||||
| BC034692.1 | AAH34692.1 | |||
| BC070066.1 | AAH70066.1 | |||
| CR936650.1 | CAI56788.1 |
More Information
Set Membership: |
|

Genomic Map