Assay Details
Target Gene Details
Entrez Gene ID: | 64066 |
Gene Name: | matrix metallopeptidase 27 |
Gene Aliases: |
MMP-27 |
Location: |
Chr.11:102690943-102705785 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MMP27 | NM_022122.2 | NP_071405.2 | ||
| XM_011542948.2 | XP_011541250.1 | |||
| XM_011542949.2 | XP_011541251.1 | |||
| XM_011542950.2 | XP_011541252.1 | |||
| XM_017018120.1 | XP_016873609.1 | |||
| AF195192.1 | AAG28453.1 | |||
| AY358752.1 | AAQ89112.1 | |||
| BX102293.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2660550 | Chr.11:102695966 - 102698742 on Build GRCh38 | Deletion |
|
| nsv832252 | Chr.11:102574748 - 102757148 on Build GRCh38 | Gain |
|
| esv3627535 | Chr.11:102695893 - 102698786 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map