Assay Details
Target Gene Details
Entrez Gene ID: | 1211 |
Gene Name: | clathrin light chain A |
Gene Aliases: |
LCA |
Location: |
Chr.9:36190856-36212062 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1017711 | Chr.9:36073424 - 36548245 on Build GRCh38 | Gain |
|
| esv34080 | Chr.9:36005887 - 36453179 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map