Assay Details
Target Gene Details
Entrez Gene ID: | 96764 |
Gene Name: | trimethylguanosine synthase 1 |
Gene Aliases: |
NCOA6IP, PIMT, PIPMT |
Location: |
Chr.8:55773232-55826445 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGS1 | NM_001317902.1 | 3 | 1449 | NP_001304831.1 |
| NM_024831.7 | 4 | 1514 | NP_079107.6 | |
| XM_005251328.2 | 3 | 1229 | XP_005251385.1 | |
| XM_006716485.2 | 4 | 1300 | XP_006716548.1 | |
| XM_017014004.1 | 4 | 1300 | XP_016869493.1 | |
| AF286340.1 | 4 | 1604 | AAK83025.1 | |
| AF432215.1 | 4 | 1262 | AAL99922.1 | |
| AK055178.1 | 1 | 388 | ||
| AK314693.1 | 4 | 1288 | ||
| AY028423.1 | 4 | 1298 | AAK27730.1 | |
| AY534911.1 | 1 | 304 | AAT02709.1 | |
| BX648548.1 | 3 | 1212 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv459n21 | Chr.8:55676869 - 56030580 on Build GRCh38 | Gain |
|
| nsv527341 | Chr.8:55679732 - 55895699 on Build GRCh38 | Gain |
|
| nsv611393 | Chr.8:55763923 - 55993509 on Build GRCh38 | Gain |
|
| esv2422388 | Chr.8:55707094 - 55812437 on Build GRCh38 | Duplication |
|
| nsv611392 | Chr.8:55741553 - 55878302 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map