Assay Details
Target Gene Details
Entrez Gene ID: | 83902 |
Gene Name: | keratin associated protein 17-1 |
Gene Aliases: |
KAP17.1, KRTAP16.1, KRTAP17.1 |
Location: |
Chr.17:41314917-41315695 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRTAP17-1 | NM_031964.1 | 1 | 544 | NP_114170.1 |
| AJ406952.1 | 1 | 544 | CAC27591.1 | |
| BC069078.1 | 1 | 542 | AAH69078.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055426 | Chr.17:41266750 - 41322116 on Build GRCh38 | Gain |
|
| esv2758690 | Chr.17:41145123 - 41740591 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79468542] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map