Assay Details
Target Gene Details
Entrez Gene ID: | 1490 |
Gene Name: | connective tissue growth factor |
Gene Aliases: |
CCN2, HCS24, IGFBP8, NOV2 |
Location: |
Chr.6:131948176-131951378 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTGF | NM_001901.2 | 5 | 2278 | NP_001892.1 |
| AK092280.1 | 5 | 2129 | ||
| AK125220.1 | 5 | 2060 | ||
| AY395801.1 | AAQ95223.1 | |||
| AY550024.1 | AAS55639.1 | |||
| BC087839.1 | 5 | 2254 | AAH87839.1 | |
| U14750.1 | 4 | 1852 | AAA75378.1 | |
| X78947.1 | 5 | 2217 | CAA55544.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7968 | Chr.6:131596032 - 131999959 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map