Assay Details
Target Gene Details
Entrez Gene ID: | 7430 |
Gene Name: | ezrin |
Gene Aliases: |
CVIL, CVL, HEL-S-105, VIL2 |
Location: |
Chr.6:158765741-158819424 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EZR | NM_001111077.1 | 14 | 2836 | NP_001104547.1 |
| NM_003379.4 | 13 | 2870 | NP_003370.2 | |
| XM_011536110.1 | 10 | 2247 | XP_011534412.1 | |
| AI366687.1 | 1 | 303 | ||
| AK129727.1 | 7 | 1792 | ||
| AL162086.1 | 14 | 2788 | CAB82418.1 | |
| BC013903.2 | 13 | 2787 | AAH13903.1 | |
| BC068458.1 | 13 | 2796 | AAH68458.1 | |
| FJ224318.1 | 13 | 2772 | ||
| J05021.1 | 13 | 2731 | AAA61278.1 | |
| X51521.1 | 13 | 2772 | CAA35893.1 |
Target Gene Details
Entrez Gene ID: | 94120 |
Gene Name: | synaptotagmin like 3 |
Gene Aliases: |
SLP3 |
Location: |
Chr.6:158644881-158766298 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYTL3 | XM_005267215.4 | 18 | 4138 | XP_005267272.1 |
| XM_005267218.4 | 18 | 3449 | XP_005267275.1 | |
| XM_005267222.4 | 10 | 2720 | XP_005267279.1 | |
| XM_006715605.3 | 19 | 4230 | XP_006715668.1 | |
| XM_006715606.3 | 19 | 4151 | XP_006715669.1 | |
| XM_006715611.3 | 18 | 3753 | XP_006715674.1 | |
| XM_011536254.2 | 19 | 3919 | XP_011534556.1 | |
| XM_011536255.2 | 9 | 2975 | XP_011534557.1 | |
| XM_017011496.1 | 18 | 4319 | XP_016866985.1 | |
| XM_017011497.1 | 19 | 3656 | XP_016866986.1 | |
| XM_017011498.1 | 15 | 3469 | XP_016866987.1 |
More Information
Set Membership: |
|

Genomic Map