Assay Details
Target Gene Details
Entrez Gene ID: | 5255 |
Gene Name: | phosphorylase kinase regulatory subunit alpha 1 |
Gene Aliases: |
PHKA |
Location: |
Chr.X:72578814-72714181 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHKA1 | NM_001122670.1 | 22 | 2745 | NP_001116142.1 |
| NM_001172436.1 | 21 | 2568 | NP_001165907.1 | |
| NM_002637.3 | 22 | 2745 | NP_002628.2 | |
| XM_006724661.2 | 21 | 2410 | XP_006724724.1 | |
| BC104944.1 | 22 | 2491 | AAI04945.1 | |
| BC143499.1 | 22 | 2491 | ||
| BC143501.1 | 21 | 2314 | ||
| X73874.1 | 22 | 2605 | CAA52083.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2663199 | Chr.X:72589543 - 72671051 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map