Assay Details
Target Gene Details
Entrez Gene ID: | 57820 |
Gene Name: | cyclin B1 interacting protein 1 |
Gene Aliases: |
C14orf18, HEI10 |
Location: |
Chr.14:20311368-20333312 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNB1IP1 | NM_021178.4 | 2 | 104 | NP_067001.3 |
| NM_182849.2 | NP_878269.1 | |||
| NM_182852.3 | 2 | 104 | NP_878272.1 | |
| AK026233.1 | 2 | 71 | BAB15403.1 | |
| AK225253.1 | 2 | 76 | ||
| AK225451.1 | 2 | 71 | ||
| AK226037.1 | 2 | 76 | ||
| AK309183.1 | 2 | 73 | ||
| AL161994.1 | 2 | 84 | CAB82326.1 | |
| BC000369.2 | 1 | 46 | AAH00369.1 | |
| BC001218.1 | 1 | 49 | AAH01218.1 | |
| BC004435.1 | 1 | 46 | AAH04435.1 | |
| BI763317.1 | 1 | 51 | ||
| BI771433.1 | 2 | 88 | ||
| BI917081.1 | 2 | 95 | ||
| BM741413.1 | ||||
| BX161423.1 | 1 | 49 | CAD61896.1 | |
| BX161424.1 | 1 | 52 | CAD61897.1 | |
| BX247978.1 | 2 | 212 | CAD62312.1 | |
| BX334446.2 | 1 | 46 | ||
| CB113193.1 | 2 | 86 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1042025 | Chr.14:20319802 - 20349692 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map