Assay Details
Target Gene Details
Entrez Gene ID: | 131566 |
Gene Name: | discoidin, CUB and LCCL domain containing 2 |
Gene Aliases: |
CLCP1, ESDN |
Location: |
Chr.3:98795970-98901689 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCBLD2 | NM_080927.3 | 16 | 4078 | NP_563615.3 |
| XM_011512419.2 | 15 | 3794 | XP_011510721.1 | |
| AB073146.1 | 16 | 4078 | BAB91138.1 | |
| AF104313.1 | 1 | 623 | ||
| AK001362.1 | 1 | 1080 | ||
| BC029658.1 | 16 | 3639 | AAH29658.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv591065 | Chr.3:98485354 - 99047482 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map