Assay Details
Target Gene Details
Entrez Gene ID: | 5516 |
Gene Name: | protein phosphatase 2 catalytic subunit beta |
Gene Aliases: |
PP2Abeta, PP2CB |
Location: |
Chr.8:30785610-30812836 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP2CB | NM_001009552.1 | 1 | 483 | NP_001009552.1 |
| AF444006.1 | 1 | 69 | AAL35904.1 | |
| AK091875.1 | 2 | 247 | ||
| BC012022.1 | 1 | 94 | AAH12022.1 | |
| BG716837.1 | 1 | 520 | ||
| BT019526.1 | 1 | 68 | AAV38333.1 | |
| CR541747.1 | 1 | 68 | CAG46547.1 | |
| J03805.1 | 1 | 66 | AAB38020.1 | |
| X12656.1 | 1 | 89 | CAA31183.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1077034 | Chr.8:30812183 - 30812884 on Build GRCh38 | Deletion |
|
| nsv1144225 | Chr.8:30812284 - 30812684 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map