Assay Details
Target Gene Details
Entrez Gene ID: | 24138 |
Gene Name: | interferon induced protein with tetratricopeptide repeats 5 |
Gene Aliases: |
ISG58, P58, RI58 |
Location: |
Chr.10:89414568-89421002 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFIT5 | NM_012420.2 | NP_036552.1 | ||
| U34605.1 | AAA84934.1 |
More Information
Set Membership: |
|

Genomic Map