Assay Details
Target Gene Details
Entrez Gene ID: | 54788 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member B12 |
Gene Aliases: |
DJ10 |
Location: |
Chr.10:72332830-72355149 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJB12 | NM_001002762.2 | 8 | 2566 | NP_001002762.2 |
| NM_017626.4 | NP_060096.3 | |||
| XM_005269931.1 | XP_005269988.1 | |||
| XM_005269932.3 | XP_005269989.1 | |||
| AK000034.1 | BAA90896.1 | |||
| AK025942.1 | 6 | 2015 | BAB15289.1 | |
| AK027259.1 | ||||
| AK123129.1 | 3 | 2502 | ||
| AK225051.1 | ||||
| AK302071.1 | ||||
| AL050381.1 | 2 | 1370 | CAH56413.1 | |
| AL833358.1 | 1 | 635 | ||
| BC011812.2 | 6 | 1935 | AAH11812.2 | |
| BC064920.1 | AAH64920.1 |

Genomic Map